Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGAAGATGGCGAGCTGCACGTC[C/T]GTGATCATCCTGCGGAGAAGAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615900 MIM: 130660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGBL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGBL5 - ATP/GTP binding protein like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035507.2 | 109 | Intron | NP_001030584.1 | |||
NM_021831.5 | 109 | Intron | NP_068603.4 | |||
XM_005264477.2 | 109 | Intron | XP_005264534.1 | |||
XM_011533011.2 | 109 | Intron | XP_011531313.1 | |||
XM_011533012.2 | 109 | Intron | XP_011531314.1 | |||
XM_011533013.2 | 109 | Intron | XP_011531315.1 |
EMILIN1 - elastin microfibril interfacer 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OST4 - oligosaccharyltransferase complex subunit 4, non-catalytic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134693.1 | 109 | Silent Mutation | ACA,ACG | T,T 3 | NP_001128165.1 |