Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGATGTGAACAGCCCAGTCAGAC[A/G]TCAACAGCTGAGGGATTGCCACACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXorf38 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXorf38 - chromosome X open reading frame 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144970.2 | 676 | Missense Mutation | ACG,ATG | T,M 210 | NP_659407.1 | |
XM_005272589.2 | 676 | Missense Mutation | ACG,ATG | T,M 210 | XP_005272646.2 | |
XM_006724527.3 | 676 | Intron | XP_006724590.2 | |||
XM_006724528.2 | 676 | Missense Mutation | ACG,ATG | T,M 165 | XP_006724591.1 | |
XM_017029302.1 | 676 | Missense Mutation | ACG,ATG | T,M 221 | XP_016884791.1 | |
XM_017029303.1 | 676 | Missense Mutation | ACG,ATG | T,M 165 | XP_016884792.1 | |
XM_017029304.1 | 676 | Missense Mutation | ACG,ATG | T,M 91 | XP_016884793.1 | |
XM_017029305.1 | 676 | Missense Mutation | ACG,ATG | T,M 91 | XP_016884794.1 |