Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAATTTAAGTTCGTGTGGTTTTA[C/T]CTTTTCCGGGAGTCTCCAGCTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191760 MIM: 613580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC647115 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC647115 - translation initiation factor IF-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UGP2 - UDP-glucose pyrophosphorylase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001521.1 | 9 | Intron | NP_001001521.1 | |||
NM_006759.3 | 9 | UTR 5 | NP_006750.3 | |||
XM_005264537.2 | 9 | Intron | XP_005264594.1 | |||
XM_005264538.1 | 9 | Intron | XP_005264595.1 | |||
XM_006712088.1 | 9 | Intron | XP_006712151.1 | |||
XM_011533091.1 | 9 | Intron | XP_011531393.1 | |||
XM_017004855.1 | 9 | Intron | XP_016860344.1 | |||
XM_017004856.1 | 9 | Intron | XP_016860345.1 | |||
XM_017004857.1 | 9 | Intron | XP_016860346.1 |
WDPCP - WD repeat containing planar cell polarity effector | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |