Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613201 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHTF18 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHTF18 - chromosome transmission fidelity factor 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPUSD1 - RNA pseudouridylate synthase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324086.1 | 785 | Missense Mutation | CCG,CTG | P,L 250 | NP_001311015.1 | |
NM_001324410.1 | 785 | Missense Mutation | CCG,CTG | P,L 121 | NP_001311339.1 | |
NM_001324411.1 | 785 | UTR 3 | NP_001311340.1 | |||
NM_001324412.1 | 785 | Missense Mutation | CCG,CTG | P,L 118 | NP_001311341.1 | |
NM_001324413.1 | 785 | Missense Mutation | CCG,CTG | P,L 118 | NP_001311342.1 | |
NM_001324414.1 | 785 | Missense Mutation | CCG,CTG | P,L 118 | NP_001311343.1 | |
NM_001324415.1 | 785 | Missense Mutation | CCG,CTG | P,L 118 | NP_001311344.1 | |
NM_058192.2 | 785 | Missense Mutation | CCG,CTG | P,L 247 | NP_478072.1 |
Set Membership: |
HapMap |