Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTCATATTAACATATTAAATCA[A/G]CACACATTTACTAAGCACTGCATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611176 MIM: 614811 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JKAMP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JKAMP - JNK1/MAPK8-associated membrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098625.2 | Intron | NP_001092095.1 | ||||
NM_001284201.1 | Intron | NP_001271130.1 | ||||
NM_001284202.1 | Intron | NP_001271131.1 | ||||
NM_001284203.1 | Intron | NP_001271132.1 | ||||
NM_001284204.1 | Intron | NP_001271133.1 | ||||
NM_016475.4 | Intron | NP_057559.2 | ||||
XM_006720171.3 | Intron | XP_006720234.1 | ||||
XM_017021366.1 | Intron | XP_016876855.1 |
L3HYPDH - trans-L-3-hydroxyproline dehydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144581.1 | Intron | NP_653182.1 |