Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGAAGGATGGAGAGAAGGAACTG[G/T]AGCTTAAAAGGGGGTGAAGTTCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604672 | ||||||||||||||||||||
Literature Links: |
CD209 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD209 - CD209 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144893.1 | 1175 | UTR 3 | NP_001138365.1 | |||
NM_001144894.1 | 1175 | UTR 3 | NP_001138366.1 | |||
NM_001144895.1 | 1175 | UTR 3 | NP_001138367.1 | |||
NM_001144896.1 | 1175 | UTR 3 | NP_001138368.1 | |||
NM_001144897.1 | 1175 | UTR 3 | NP_001138369.1 | |||
NM_001144899.1 | 1175 | UTR 3 | NP_001138371.1 | |||
NM_021155.3 | 1175 | UTR 3 | NP_066978.1 |