Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTACGGGGGTGGCAGGCAATCAGC[A/G]CATGCTCCATGCAGTTGGTCACCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RDH16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RDH16 - retinol dehydrogenase 16 (all-trans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320108.1 | 2724 | Missense Mutation | GCG,GTG | A,V 126 | NP_001307037.1 | |
NM_003708.4 | 2724 | Missense Mutation | GCG,GTG | A,V 271 | NP_003699.3 |