Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAACTCATAAAATGCTTGCTTACA[A/G]CCCTACGACAAAGAAGGCTCCCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164176 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEDD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DEDD2 - death effector domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270614.1 | Intron | NP_001257543.1 | ||||
NM_001270615.1 | Intron | NP_001257544.1 | ||||
NM_133328.3 | Intron | NP_579874.1 | ||||
XM_011526569.1 | Intron | XP_011524871.1 | ||||
XM_011526571.1 | Intron | XP_011524873.1 | ||||
XM_011526572.1 | Intron | XP_011524874.1 | ||||
XM_017026402.1 | Intron | XP_016881891.1 | ||||
XM_017026403.1 | Intron | XP_016881892.1 |
POU2F2 - POU class 2 homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |