Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCGCGCTGTGCGCCCCGCGGA[A/G]CCCGCGAACCCCGCTCGCTGCCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608488 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMOC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMOC1 - SPARC related modular calcium binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034852.2 | 237 | UTR 5 | NP_001030024.1 | |||
NM_022137.5 | 237 | UTR 5 | NP_071420.1 | |||
XM_005267995.1 | 237 | UTR 5 | XP_005268052.1 | |||
XM_005267996.1 | 237 | UTR 5 | XP_005268053.1 |