Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGGCTACAAGCTGAGGCCTGATC[A/G]GGAATTCAGAGTTGGCATTCAGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 108728 | ||||||||||||||||||||
Literature Links: |
ACLY PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACLY - ATP citrate lyase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC25 - tetratricopeptide repeat domain 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031421.3 | 442 | Missense Mutation | CAG,CGG | Q,R 114 | NP_113609.1 | |
XM_011525323.2 | 442 | Intron | XP_011523625.1 | |||
XM_011525324.2 | 442 | Missense Mutation | CAG,CGG | Q,R 114 | XP_011523626.1 |