Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGAAGTCAGCTCCCGCGAGGCA[A/G]CCGGTCTCGGGGCTCCGCGGGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610339 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf50 - chromosome 1 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024097.3 | Intron | NP_077002.2 |
LOC105378683 - uncharacterized LOC105378683 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
P3H1 - prolyl 3-hydroxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146289.1 | Intron | NP_001139761.1 | ||||
NM_001243246.1 | Intron | NP_001230175.1 | ||||
NM_022356.3 | Intron | NP_071751.3 | ||||
XM_005271110.2 | Intron | XP_005271167.1 | ||||
XM_011541948.1 | Intron | XP_011540250.1 | ||||
XM_017002051.1 | Intron | XP_016857540.1 | ||||
XM_017002052.1 | Intron | XP_016857541.1 |