Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAATGCAAGATCCCAGCTGTAT[G/T]CTGGTGATAACAGCTAGGTGGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613699 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLT6D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLT6D1 - glycosyltransferase 6 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182974.2 | 1423 | UTR 3 | NP_892019.2 | |||
XM_011518633.1 | 1423 | UTR 3 | XP_011516935.1 | |||
XM_011518634.1 | 1423 | UTR 3 | XP_011516936.1 | |||
XM_011518635.1 | 1423 | UTR 3 | XP_011516937.1 | |||
XM_011518636.1 | 1423 | UTR 3 | XP_011516938.1 | |||
XM_011518637.1 | 1423 | UTR 3 | XP_011516939.1 | |||
XM_011518638.2 | 1423 | UTR 3 | XP_011516940.1 |
LOC102723971 - lipocalin 1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |