Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCTCTCATTGATGATTATTTCC[A/G]TGGAGTGGACGCATGGTGTCTTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612483 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAT3 - FAT atypical cadherin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008781.2 | Intron | NP_001008781.2 | ||||
XM_017017178.1 | Intron | XP_016872667.1 | ||||
XM_017017179.1 | Intron | XP_016872668.1 | ||||
XM_017017180.1 | Intron | XP_016872669.1 | ||||
XM_017017181.1 | Intron | XP_016872670.1 | ||||
XM_017017182.1 | Intron | XP_016872671.1 | ||||
XM_017017183.1 | Intron | XP_016872672.1 | ||||
XM_017017184.1 | Intron | XP_016872673.1 | ||||
XM_017017185.1 | Intron | XP_016872674.1 | ||||
XM_017017186.1 | Intron | XP_016872675.1 | ||||
XM_017017187.1 | Intron | XP_016872676.1 |