Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGATGGAAGTGGACCATGACCG[G/T]CAGGTGGTGCATGTGGAGACACTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615126 MIM: 109635 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD13D PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKRD13D - ankyrin repeat domain 13D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207354.2 | 12 | Silent Mutation | CGG,CGT | R,R 193 | NP_997237.2 | |
XM_011544985.1 | 12 | Silent Mutation | CGG,CGT | R,R 193 | XP_011543287.1 | |
XM_011544986.1 | 12 | Silent Mutation | CGG,CGT | R,R 106 | XP_011543288.1 | |
XM_011544987.2 | 12 | Silent Mutation | CGG,CGT | R,R 106 | XP_011543289.1 | |
XM_011544988.2 | 12 | UTR 5 | XP_011543290.1 |
GRK2 - G protein-coupled receptor kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |