Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HDX - highly divergent homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177478.1 | 1783 | Silent Mutation | CCA,CCG | P,P 503 | NP_001170949.1 | |
NM_001177479.1 | 1783 | Silent Mutation | CCA,CCG | P,P 561 | NP_001170950.1 | |
NM_144657.4 | 1783 | Silent Mutation | CCA,CCG | P,P 561 | NP_653258.2 | |
XM_006724619.1 | 1783 | Silent Mutation | CCA,CCG | P,P 503 | XP_006724682.1 | |
XM_011530863.2 | 1783 | Silent Mutation | CCA,CCG | P,P 561 | XP_011529165.1 | |
XM_011530864.1 | 1783 | Silent Mutation | CCA,CCG | P,P 561 | XP_011529166.1 | |
XM_017029274.1 | 1783 | Silent Mutation | CCA,CCG | P,P 561 | XP_016884763.1 | |
XM_017029275.1 | 1783 | Silent Mutation | CCA,CCG | P,P 543 | XP_016884764.1 |