Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTTGGCCTCCAAGTCTTCAAGG[A/C]AGAGCCCATGAACTGCAGGCAGTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611335 MIM: 611325 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNORA5A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SNORA5A - small nucleolar RNA, H/ACA box 5A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA5B - small nucleolar RNA, H/ACA box 5B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA5C - small nucleolar RNA, H/ACA box 5C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBRG4 - transforming growth factor beta regulator 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261834.1 | Intron | NP_001248763.1 | ||||
NM_004749.3 | Intron | NP_004740.2 | ||||
NM_030900.3 | Intron | NP_112162.1 | ||||
NM_199122.2 | Intron | NP_954573.1 |