Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGACGTGCAGGACCTCCTGAAGG[A/G]TCTGAAGGCTCAGTCGCAGGTCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM26F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM26F - family with sequence similarity 26 member F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010919.2 | 908 | Missense Mutation | GAT,GGT | D,G 169 | NP_001010919.1 | |
NM_001276460.1 | 908 | Intron | NP_001263389.1 | |||
XM_005266999.3 | 908 | Missense Mutation | GAT,GGT | D,G 169 | XP_005267056.1 | |
XM_011535845.2 | 908 | Missense Mutation | GAT,GGT | D,G 169 | XP_011534147.1 |