Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGATGTAATTGAAGAATTCAGTT[C/G]CATTCCTTCCACAACATCTGAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153619 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLGAP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLGAP5 - DLG associated protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146015.1 | 2474 | Missense Mutation | CAA,GAA | Q,E 753 | NP_001139487.1 | |
NM_014750.4 | 2474 | Missense Mutation | CAA,GAA | Q,E 753 | NP_055565.3 | |
XM_017021840.1 | 2474 | Missense Mutation | CAA,GAA | Q,E 753 | XP_016877329.1 |
LGALS3 - lectin, galactoside binding soluble 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |