Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCACGTGGATGGGCCCCCCTGGC[A/G]TCAGGAGCCCGTGGGTTTTTGCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP27C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP27C1 - cytochrome P450 family 27 subfamily C member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001665.3 | 1207 | Missense Mutation | ACG,ATG | T,M 359 | NP_001001665.3 | |
XM_017003960.1 | 1207 | Missense Mutation | ACG,ATG | T,M 456 | XP_016859449.1 | |
XM_017003961.1 | 1207 | Intron | XP_016859450.1 |