Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGATGAGATTGAAATTGAGCCA[C/G]TACGACAGAGCAGCGATAAAATGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC74A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC74A - leucine rich repeat containing 74A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194287.2 | 218 | Missense Mutation | CTA,GTA | L,V 37 | NP_919263.2 | |
XM_011536479.1 | 218 | UTR 5 | XP_011534781.1 | |||
XM_011536483.1 | 218 | UTR 5 | XP_011534785.1 | |||
XM_011536487.1 | 218 | UTR 5 | XP_011534789.1 | |||
XM_017021032.1 | 218 | UTR 5 | XP_016876521.1 | |||
XM_017021033.1 | 218 | UTR 5 | XP_016876522.1 | |||
XM_017021034.1 | 218 | Missense Mutation | CTA,GTA | L,V 37 | XP_016876523.1 | |
XM_017021035.1 | 218 | Missense Mutation | CTA,GTA | L,V 20 | XP_016876524.1 | |
XM_017021036.1 | 218 | UTR 5 | XP_016876525.1 | |||
XM_017021037.1 | 218 | UTR 5 | XP_016876526.1 | |||
XM_017021038.1 | 218 | UTR 5 | XP_016876527.1 | |||
XM_017021039.1 | 218 | Intron | XP_016876528.1 | |||
XM_017021040.1 | 218 | Intron | XP_016876529.1 | |||
XM_017021041.1 | 218 | Intron | XP_016876530.1 |