Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGTGTTGGCGCTTGTTGCATTC[A/G]TTGCTGCTCCAAGTTAAAAAGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609846 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
REG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
REG4 - regenerating family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159352.1 | 698 | Silent Mutation | AAC,AAT | N,N 144 | NP_001152824.1 | |
NM_001159353.1 | 698 | Intron | NP_001152825.1 | |||
NM_032044.3 | 698 | Silent Mutation | AAC,AAT | N,N 144 | NP_114433.1 |