Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCATCTGGCCAGCATGCTGCTCT[A/G]CACGGCTCGCCTGGTCGGCCTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606225 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOL9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOL9 - nucleolar protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024654.4 | 107 | Intron | NP_078930.3 | |||
XM_005263493.3 | 107 | Intron | XP_005263550.1 | |||
XM_011542147.2 | 107 | Intron | XP_011540449.1 | |||
XM_017002337.1 | 107 | Intron | XP_016857826.1 | |||
XM_017002338.1 | 107 | Intron | XP_016857827.1 | |||
XM_017002339.1 | 107 | Intron | XP_016857828.1 |
TAS1R1 - taste 1 receptor member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138697.3 | 107 | Missense Mutation | TAC,TGC | Y,C 4 | NP_619642.2 | |
NM_177540.2 | 107 | Missense Mutation | TAC,TGC | Y,C 4 | NP_803884.1 | |
XM_011542203.2 | 107 | Intron | XP_011540505.1 | |||
XM_011542206.2 | 107 | Intron | XP_011540508.1 | |||
XM_017002402.1 | 107 | Intron | XP_016857891.1 | |||
XM_017002403.1 | 107 | Intron | XP_016857892.1 |