Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGCTCTACAAGATCGTGCGCGG[A/G]GACCAGCCGGCGGCCAGCGGCGACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300435 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107985692 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC107985692 - A-kinase anchor protein 17B-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PGRMC1 - progesterone receptor membrane component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282621.1 | 263 | Silent Mutation | GGA,GGG | G,G 48 | NP_001269550.1 | |
NM_006667.4 | 263 | Silent Mutation | GGA,GGG | G,G 48 | NP_006658.1 |
Set Membership: |
HapMap |