Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTCCTTGATACACTTGCTCAGA[C/T]CCCTCGGCTACTCTCTTGCCCAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609925 MIM: 609926 | ||||||||||||||||||||
Literature Links: |
DPEP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPEP2 - dipeptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324159.1 | Intron | NP_001311088.1 | ||||
NM_022355.3 | Intron | NP_071750.1 | ||||
XM_005256090.4 | Intron | XP_005256147.2 | ||||
XM_011523266.1 | Intron | XP_011521568.1 | ||||
XM_011523268.2 | Intron | XP_011521570.1 | ||||
XM_011523271.2 | Intron | XP_011521573.1 | ||||
XM_011523272.2 | Intron | XP_011521574.1 | ||||
XM_011523273.2 | Intron | XP_011521575.1 | ||||
XM_011523274.2 | Intron | XP_011521576.1 | ||||
XM_017023541.1 | Intron | XP_016879030.1 | ||||
XM_017023542.1 | Intron | XP_016879031.1 | ||||
XM_017023543.1 | Intron | XP_016879032.1 | ||||
XM_017023544.1 | Intron | XP_016879033.1 | ||||
XM_017023545.1 | Intron | XP_016879034.1 | ||||
XM_017023546.1 | Intron | XP_016879035.1 | ||||
XM_017023547.1 | Intron | XP_016879036.1 |
DPEP3 - dipeptidase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |