Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGTGCATGAGTCCATTTTCTTT[A/T]AACTCTGAAAGAATAAAGTGGTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615652 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACOT13 - acyl-CoA thioesterase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160094.1 | 1625 | Intron | NP_001153566.1 | |||
NM_018473.3 | 1625 | Intron | NP_060943.1 |
C6orf62 - chromosome 6 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030939.4 | 1625 | UTR 3 | NP_112201.1 | |||
XM_005249433.3 | 1625 | Intron | XP_005249490.1 | |||
XM_011514928.2 | 1625 | UTR 3 | XP_011513230.1 |