Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGTCACAGGATGCCTTTTAAGCA[A/G]ATCACTAATAGAAAACAATTTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDX - highly divergent homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177478.1 | Intron | NP_001170949.1 | ||||
NM_001177479.1 | Intron | NP_001170950.1 | ||||
NM_144657.4 | Intron | NP_653258.2 | ||||
XM_006724619.1 | Intron | XP_006724682.1 | ||||
XM_011530863.2 | Intron | XP_011529165.1 | ||||
XM_011530864.1 | Intron | XP_011529166.1 | ||||
XM_017029274.1 | Intron | XP_016884763.1 | ||||
XM_017029275.1 | Intron | XP_016884764.1 |