Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAGGCCTGGTCGATTTTCTGAGG[A/G]CTCAGGGACAGGTTGTAAATGAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf87 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf87 - chromosome 1 open reading frame 87 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152377.2 | 1469 | Silent Mutation | AGC,AGT | S,S 517 | NP_689590.1 | |
XM_017000307.1 | 1469 | Silent Mutation | AGC,AGT | S,S 469 | XP_016855796.1 | |
XM_017000308.1 | 1469 | Intron | XP_016855797.1 |