Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGTCAGACAGCAACACTGACCA[C/T]GTGAGTAGGTAACATTTCTAATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608486 MIM: 601445 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTSS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTSS1 - metastasis suppressor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFB9 - NADH:ubiquinone oxidoreductase subunit B9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278645.1 | Intron | NP_001265574.1 | ||||
NM_001278646.1 | Intron | NP_001265575.1 | ||||
NM_001311168.1 | Intron | NP_001298097.1 | ||||
NM_005005.2 | Intron | NP_004996.1 |
TATDN1 - TatD DNase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |