Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACACCATCTGAATTGCCTGCAAA[C/G]CCAGGTAAGAAGCTGGGCTCTACAG
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 605659 | |||||||||||||||||||||||||||||
Literature Links: |
CLEC2D PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CLEC2D - C-type lectin domain family 2 member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004419.4 | 84 | Silent Mutation | AAC,AAG | N,K 19 | NP_001004419.1 | |
NM_001197317.2 | 84 | Silent Mutation | AAC,AAG | N,K 19 | NP_001184246.1 | |
NM_001197318.2 | 84 | Silent Mutation | AAC,AAG | N,K 19 | NP_001184247.1 | |
NM_001197319.2 | 84 | Silent Mutation | AAC,AAG | N,K 19 | NP_001184248.1 | |
NM_013269.5 | 84 | Silent Mutation | AAC,AAG | N,K 19 | NP_037401.1 |