Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616142 MIM: 603962 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM98B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM98B - family with sequence similarity 98 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASGRP1 - RAS guanyl releasing protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128602.1 | Intron | NP_001122074.1 | ||||
NM_001306086.1 | Intron | NP_001293015.1 | ||||
NM_005739.3 | Intron | NP_005730.2 | ||||
XM_005254114.2 | Intron | XP_005254171.1 | ||||
XM_011521151.2 | Intron | XP_011519453.1 | ||||
XM_017021860.1 | Intron | XP_016877349.1 |
Set Membership: |
HapMap |