Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCAGCACCTGGGCCATGTAGCT[C/T]TCCTGCTGCTTCTGGATCCGGAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609447 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MZB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MZB1 - marginal zone B and B1 cell specific protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PROB1 - proline rich basic protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA24 - spermatogenesis associated 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194296.1 | 555 | Silent Mutation | GAA,GAG | E,E 176 | NP_919272.1 | |
XM_005271916.4 | 555 | Missense Mutation | AAA,AGA | K,R 185 | XP_005271973.1 | |
XM_011543252.2 | 555 | Missense Mutation | AAA,AGA | K,R 146 | XP_011541554.1 | |
XM_011543253.2 | 555 | Missense Mutation | AAA,AGA | K,R 146 | XP_011541555.1 | |
XM_011543254.2 | 555 | Intron | XP_011541556.1 |