Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615692 MIM: 602644 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHID1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHID1 - chitinase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142674.1 | 1231 | Missense Mutation | NP_001136146.1 | |||
NM_001142675.1 | 1231 | Missense Mutation | NP_001136147.1 | |||
NM_001142676.1 | 1231 | Missense Mutation | NP_001136148.1 | |||
NM_001142677.1 | 1231 | Missense Mutation | NP_001136149.1 | |||
NM_023947.3 | 1231 | Missense Mutation | NP_076436.3 | |||
XM_005253072.2 | 1231 | Missense Mutation | XP_005253129.1 | |||
XM_005253073.4 | 1231 | Missense Mutation | XP_005253130.1 | |||
XM_006718282.2 | 1231 | Missense Mutation | XP_006718345.1 | |||
XM_011520299.1 | 1231 | Missense Mutation | XP_011518601.1 | |||
XM_011520300.1 | 1231 | Missense Mutation | XP_011518602.1 | |||
XM_011520301.2 | 1231 | Intron | XP_011518603.1 | |||
XM_011520302.2 | 1231 | Intron | XP_011518604.1 | |||
XM_017018166.1 | 1231 | Missense Mutation | XP_016873655.1 | |||
XM_017018167.1 | 1231 | Intron | XP_016873656.1 | |||
XM_017018168.1 | 1231 | Intron | XP_016873657.1 |
TSPAN4 - tetraspanin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |