Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614697 MIM: 603567 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KATNAL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KATNAL2 - katanin catalytic subunit A1 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031303.2 | 120 | Intron | NP_112593.2 | |||
XM_005258357.4 | 120 | UTR 5 | XP_005258414.1 | |||
XM_005258358.4 | 120 | UTR 5 | XP_005258415.1 | |||
XM_005258361.3 | 120 | Intron | XP_005258418.1 | |||
XM_006722554.3 | 120 | UTR 5 | XP_006722617.1 | |||
XM_011526219.2 | 120 | UTR 5 | XP_011524521.1 | |||
XM_011526220.1 | 120 | UTR 5 | XP_011524522.1 | |||
XM_011526221.2 | 120 | UTR 5 | XP_011524523.1 | |||
XM_011526223.2 | 120 | UTR 5 | XP_011524525.1 | |||
XM_017026028.1 | 120 | UTR 5 | XP_016881517.1 | |||
XM_017026029.1 | 120 | UTR 5 | XP_016881518.1 | |||
XM_017026030.1 | 120 | UTR 5 | XP_016881519.1 | |||
XM_017026031.1 | 120 | Intron | XP_016881520.1 | |||
XM_017026032.1 | 120 | UTR 5 | XP_016881521.1 |
Set Membership: |
HapMap |