Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCACCTGCGGGCTGCAGTCAG[C/T]CCGGACGCCGAGACCCGTGCCTAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102560 MIM: 607643 | ||||||||||||||||||||
Literature Links: |
ACTG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTG1 - actin gamma 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FSCN2 - fascin actin-bundling protein 2, retinal | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077182.2 | 1098 | Intron | NP_001070650.1 | |||
NM_012418.3 | 1098 | Intron | NP_036550.1 | |||
XM_011524587.2 | 1098 | Intron | XP_011522889.1 | |||
XM_011524590.2 | 1098 | UTR 5 | XP_011522892.1 |