Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608755 | ||||||||||||||||||||||||||||||||
Literature Links: |
LLGL2 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
LLGL2 - LLGL2, scribble cell polarity complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015002.1 | Intron | NP_001015002.1 | ||||
NM_001031803.1 | Intron | NP_001026973.1 | ||||
NM_004524.2 | Intron | NP_004515.2 | ||||
XM_011524801.1 | Intron | XP_011523103.1 | ||||
XM_011524802.1 | Intron | XP_011523104.1 | ||||
XM_011524807.2 | Intron | XP_011523109.1 | ||||
XM_017024625.1 | Intron | XP_016880114.1 | ||||
XM_017024626.1 | Intron | XP_016880115.1 | ||||
XM_017024627.1 | Intron | XP_016880116.1 | ||||
XM_017024628.1 | Intron | XP_016880117.1 | ||||
XM_017024629.1 | Intron | XP_016880118.1 | ||||
XM_017024630.1 | Intron | XP_016880119.1 | ||||
XM_017024631.1 | Intron | XP_016880120.1 | ||||
XM_017024632.1 | Intron | XP_016880121.1 |
TSEN54 - tRNA splicing endonuclease subunit 54 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |