Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCGCTTGCTCAGGTTGCCCTGG[A/G]TAGTGGCCCAGATGGAGGGATGGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605475 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAIAP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BAIAP2 - BAI1 associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144888.1 | Intron | NP_001138360.1 | ||||
NM_006340.2 | Intron | NP_006331.1 | ||||
NM_017450.2 | Intron | NP_059344.1 | ||||
NM_017451.2 | Intron | NP_059345.1 | ||||
XM_005256943.1 | Intron | XP_005257000.1 | ||||
XM_005256944.1 | Intron | XP_005257001.1 | ||||
XM_005256945.1 | Intron | XP_005257002.1 | ||||
XM_005256948.3 | Intron | XP_005257005.1 | ||||
XM_006721635.1 | Intron | XP_006721698.1 | ||||
XM_006721636.1 | Intron | XP_006721699.1 | ||||
XM_006721637.2 | Intron | XP_006721700.1 | ||||
XM_011524193.1 | Intron | XP_011522495.1 | ||||
XM_011524194.1 | Intron | XP_011522496.1 | ||||
XM_011524195.1 | Intron | XP_011522497.1 | ||||
XM_011524196.1 | Intron | XP_011522498.1 | ||||
XM_011524197.1 | Intron | XP_011522499.1 | ||||
XM_017024017.1 | Intron | XP_016879506.1 | ||||
XM_017024018.1 | Intron | XP_016879507.1 | ||||
XM_017024019.1 | Intron | XP_016879508.1 |
BAIAP2-AS1 - BAIAP2 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |