Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604782 MIM: 600617 | ||||||||||||||||||||
Literature Links: |
ASH2L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASH2L - ASH2 like histone lysine methyltransferase complex subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAR - steroidogenic acute regulatory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000349.2 | 872 | Missense Mutation | GAC,GCC | D,A 203 | NP_000340.2 | |
XM_006716392.2 | 872 | Missense Mutation | GAC,GCC | D,A 203 | XP_006716455.1 |