Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATCCATGTTTTCGGTTACTCTG[C/T]AAAACAGAGGAAAAAAATTATTTAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611197 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JAKMIP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
JAKMIP2 - janus kinase and microtubule interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270934.1 | Intron | NP_001257863.1 | ||||
NM_001270941.1 | Intron | NP_001257870.1 | ||||
NM_001282282.1 | Intron | NP_001269211.1 | ||||
NM_014790.4 | Intron | NP_055605.2 | ||||
XM_017010097.1 | Intron | XP_016865586.1 | ||||
XM_017010098.1 | Intron | XP_016865587.1 |
JAKMIP2-AS1 - JAKMIP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |