Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114815 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CA8 - carbonic anhydrase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321837.1 | 5171 | Intron | NP_001308766.1 | |||
NM_001321838.1 | 5171 | Intron | NP_001308767.1 | |||
NM_001321839.1 | 5171 | UTR 3 | NP_001308768.1 | |||
NM_004056.5 | 5171 | UTR 3 | NP_004047.3 | |||
XM_011517587.2 | 5171 | Intron | XP_011515889.1 | |||
XM_011517588.2 | 5171 | Intron | XP_011515890.1 | |||
XM_017013818.1 | 5171 | Intron | XP_016869307.1 |