Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF620 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF620 - zinc finger protein 620 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256167.1 | Intron | NP_001243096.1 | ||||
NM_001256168.1 | Intron | NP_001243097.1 | ||||
NM_175888.3 | Intron | NP_787084.1 | ||||
XM_005265011.3 | Intron | XP_005265068.1 | ||||
XM_005265012.3 | Intron | XP_005265069.1 | ||||
XM_017006064.1 | Intron | XP_016861553.1 | ||||
XM_017006065.1 | Intron | XP_016861554.1 | ||||
XM_017006066.1 | Intron | XP_016861555.1 | ||||
XM_017006067.1 | Intron | XP_016861556.1 | ||||
XM_017006068.1 | Intron | XP_016861557.1 | ||||
XM_017006069.1 | Intron | XP_016861558.1 |
Set Membership: |
HapMap |