Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGGCCTATAATAATCTTGTCTC[A/G]TACTGTCCTTATCTGGTTATAATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612106 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB40 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZBTB40 - zinc finger and BTB domain containing 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083621.1 | Intron | NP_001077090.1 | ||||
NM_014870.3 | Intron | NP_055685.3 | ||||
XM_011542499.2 | Intron | XP_011540801.1 | ||||
XM_011542500.2 | Intron | XP_011540802.1 | ||||
XM_017003003.1 | Intron | XP_016858492.1 |