Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGTTTACAAGTAGTTTCTCCAG[A/G]ATCCCAGCTCTTTCCGACGCCAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
73 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602839 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3CD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIK3CD - phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005026.3 | Intron | NP_005017.3 | ||||
XM_006710687.2 | Intron | XP_006710750.1 | ||||
XM_006710689.2 | Intron | XP_006710752.1 | ||||
XM_017001476.1 | Intron | XP_016856965.1 | ||||
XM_017001477.1 | Intron | XP_016856966.1 | ||||
XM_017001478.1 | Intron | XP_016856967.1 | ||||
XM_017001479.1 | Intron | XP_016856968.1 | ||||
XM_017001480.1 | Intron | XP_016856969.1 | ||||
XM_017001481.1 | Intron | XP_016856970.1 | ||||
XM_017001482.1 | Intron | XP_016856971.1 | ||||
XM_017001483.1 | Intron | XP_016856972.1 | ||||
XM_017001484.1 | Intron | XP_016856973.1 | ||||
XM_017001485.1 | Intron | XP_016856974.1 |
PIK3CD-AS1 - PIK3CD antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |