Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
67 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613565 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE4B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UBE4B - ubiquitination factor E4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105562.2 | 241 | UTR 5 | NP_001099032.1 | |||
NM_006048.4 | 241 | UTR 5 | NP_006039.2 | |||
XM_005263422.1 | 241 | UTR 5 | XP_005263479.1 | |||
XM_011540488.2 | 241 | Intron | XP_011538790.1 | |||
XM_011540489.2 | 241 | UTR 5 | XP_011538791.1 | |||
XM_011540490.1 | 241 | UTR 5 | XP_011538792.1 | |||
XM_017000035.1 | 241 | Intron | XP_016855524.1 | |||
XM_017000036.1 | 241 | UTR 5 | XP_016855525.1 |
Set Membership: |
HapMap |