Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCCCAAGGAAGCTCTGTGAAAC[C/T]CTGGAAGGACCCCGGGCTTGCCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616886 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSE1 - Gse1 coiled-coil protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134473.2 | Intron | NP_001127945.1 | ||||
NM_001278184.1 | Intron | NP_001265113.1 | ||||
NM_014615.3 | Intron | NP_055430.1 | ||||
XM_005255859.4 | Intron | XP_005255916.3 | ||||
XM_005255860.3 | Intron | XP_005255917.3 | ||||
XM_005255861.4 | Intron | XP_005255918.3 | ||||
XM_005255863.4 | Intron | XP_005255920.1 | ||||
XM_005255864.4 | Intron | XP_005255921.1 | ||||
XM_005255865.4 | Intron | XP_005255922.1 | ||||
XM_011522965.2 | Intron | XP_011521267.1 | ||||
XM_011522966.2 | Intron | XP_011521268.1 | ||||
XM_017023083.1 | Intron | XP_016878572.1 | ||||
XM_017023084.1 | Intron | XP_016878573.1 |