Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602075 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SATB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SATB1 - SATB homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131010.3 | 5624 | UTR 3 | NP_001124482.1 | |||
NM_001195470.2 | 5624 | UTR 3 | NP_001182399.1 | |||
NM_001322871.1 | 5624 | UTR 3 | NP_001309800.1 | |||
NM_001322872.1 | 5624 | UTR 3 | NP_001309801.1 | |||
NM_001322873.1 | 5624 | UTR 3 | NP_001309802.1 | |||
NM_001322874.1 | 5624 | UTR 3 | NP_001309803.1 | |||
NM_001322875.1 | 5624 | UTR 3 | NP_001309804.1 | |||
NM_001322876.1 | 5624 | UTR 3 | NP_001309805.1 | |||
NM_002971.5 | 5624 | UTR 3 | NP_002962.1 | |||
XM_011533988.2 | 5624 | Intron | XP_011532290.1 | |||
XM_011533989.2 | 5624 | Intron | XP_011532291.1 | |||
XM_011533990.2 | 5624 | Intron | XP_011532292.1 |
Set Membership: |
HapMap |