Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608294 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR2115 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR2115 - microRNA 2115 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NME6 - NME/NM23 nucleoside diphosphate kinase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPINK8 - serine peptidase inhibitor, Kazal type 8 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080525.1 | 1256 | Intron | NP_001073994.1 | |||
XM_017007046.1 | 1256 | UTR 3 | XP_016862535.1 | |||
XM_017007047.1 | 1256 | UTR 3 | XP_016862536.1 |
Set Membership: |
HapMap |