Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCGTTTCGCTTTTGGCTTTCCCA[C/G]AGTTTCTACTGATTGTGCATCATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610853 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHCTF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AHCTF1 - AT-hook containing transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323342.1 | 6961 | Silent Mutation | CTG,GTG | L,V 2185 | NP_001310271.1 | |
NM_001323343.1 | 6961 | Silent Mutation | CTG,GTG | L,V 2185 | NP_001310272.1 | |
NM_015446.4 | 6961 | Intron | NP_056261.4 | |||
XM_006711758.1 | 6961 | Intron | XP_006711821.1 | |||
XM_011544156.2 | 6961 | Intron | XP_011542458.2 | |||
XM_011544157.2 | 6961 | Intron | XP_011542459.2 |
Set Membership: |
HapMap |