Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGCACTTTCTGAGTTGGTTAG[G/T]GCATAATAGCACAATAGGAACCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600366 | ||||||||||||||||||||
Literature Links: |
ISL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ISL1 - ISL LIM homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002202.2 | Intron | NP_002193.2 | ||||
XM_011543380.2 | Intron | XP_011541682.1 |
LOC642366 - uncharacterized LOC642366 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |