Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LYPD6B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LYPD6B - LY6/PLAUR domain containing 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317002.1 | Intron | NP_001303931.1 | ||||
NM_001317003.1 | Intron | NP_001303932.1 | ||||
NM_001317004.1 | Intron | NP_001303933.1 | ||||
NM_001317005.1 | Intron | NP_001303934.1 | ||||
NM_001317006.1 | Intron | NP_001303935.1 | ||||
NM_177964.4 | Intron | NP_808879.2 | ||||
XM_005246306.4 | Intron | XP_005246363.1 | ||||
XM_006712279.2 | Intron | XP_006712342.1 | ||||
XM_006712280.3 | Intron | XP_006712343.1 | ||||
XM_006712281.3 | Intron | XP_006712344.1 | ||||
XM_011510620.2 | Intron | XP_011508922.1 | ||||
XM_011510623.2 | Intron | XP_011508925.1 | ||||
XM_011510625.1 | Intron | XP_011508927.1 | ||||
XM_017003364.1 | Intron | XP_016858853.1 | ||||
XM_017003365.1 | Intron | XP_016858854.1 | ||||
XM_017003366.1 | Intron | XP_016858855.1 |