Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603275 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIP5K1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIP5K1A - phosphatidylinositol-4-phosphate 5-kinase type 1 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135636.1 | Intron | NP_001129108.1 | ||||
NM_001135637.1 | Intron | NP_001129109.1 | ||||
NM_001135638.1 | Intron | NP_001129110.1 | ||||
NM_003557.2 | Intron | NP_003548.1 | ||||
XM_005245525.4 | Intron | XP_005245582.1 | ||||
XM_005245527.3 | Intron | XP_005245584.1 | ||||
XM_005245528.4 | Intron | XP_005245585.1 | ||||
XM_005245530.4 | Intron | XP_005245587.1 | ||||
XM_005245531.3 | Intron | XP_005245588.1 | ||||
XM_005245532.3 | Intron | XP_005245589.1 | ||||
XM_006711563.3 | Intron | XP_006711626.1 | ||||
XM_006711564.3 | Intron | XP_006711627.1 | ||||
XM_006711567.3 | Intron | XP_006711630.1 | ||||
XM_006711568.3 | Intron | XP_006711631.2 | ||||
XM_011510043.2 | Intron | XP_011508345.1 | ||||
XM_011510045.2 | Intron | XP_011508347.1 | ||||
XM_011510046.2 | Intron | XP_011508348.1 | ||||
XM_011510048.2 | Intron | XP_011508350.1 | ||||
XM_011510049.2 | Intron | XP_011508351.1 | ||||
XM_017002439.1 | Intron | XP_016857928.1 | ||||
XM_017002440.1 | Intron | XP_016857929.1 | ||||
XM_017002441.1 | Intron | XP_016857930.1 |
Set Membership: |
HapMap |